PffBT4T-2OD (PCE11), Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66126-MC
H66126-MD
H66126-ME
AAH66126-MCEA
922.72
CAD
AAH66126-MC
AAH66126-MD
AAH66126-ME
PffBT4T-2OD (PCE11), Organic Photovoltaics (OPV) reagent
PffBT4T-2OD (PCE11)
Formula:
(C₆₂H₈₈N₂S₅F₂)n |
CAS Number:
1644164-62-4 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.63 ± 0.02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...