PTQ10, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
Poly[(thiophene)-alt-(6,7-difluoro-2-(2-hexyldecyloxy)quinoxaline)]
H66106-MC
H66106-ME
H66106-MD
AAH66106-MDEA
1205.54
CAD
AAH66106-MD
AAH66106-MC
AAH66106-ME
PTQ10, Organic Photovoltaics (OPV) reagent
PTQ10
Formula:
(C₂₈H₃₆F₂N₂OS)n |
CAS Number:
2270233-86-6 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.96 ± 0.02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...