ITIC-M, Organic Photovoltaics (OPV) reagent

Supplier: Thermo Scientific Chemicals

H66035-MC H66035-MD H66035-ME
AAH66035-MCEA 835.82 CAD
AAH66035-MC AAH66035-MD AAH66035-ME
ITIC-M, Organic Photovoltaics (OPV) reagent
ITIC-M
Formula: C₉₆H₈₆O₂N₄S₄
MW: 1456.00 g/mol
CAS Number: 2047352-80-5

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.65 ± 0.02 eV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR