PffBT4T-2DT, Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
H66014-MC
H66014-MD
H66014-ME
AAH66014-MCEA
921.14
CAD
AAH66014-MC
AAH66014-MD
AAH66014-ME
PffBT4T-2DT, Organic Photovoltaics (OPV) reagent
PffBT4T-2DT
Formula:
(C₆₆H₉₆N₂S₅F₂)n |
CAS Number:
1430201-60-7 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1.65 ± 0.02 eV |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...