VWR® Films with Colour Coded Marking Areas, Polyester
Supplier: VWR International
60941-096 60941-098 60941-100 60941-102 60941-104
60941-096CS 164.34 CAD
60941-096 60941-098 60941-100 60941-102 60941-104
VWR® Films with Colour Coded Marking Areas, Polyester
Seals Microplate Seals
Identical to polyester films above, but including a colour dot and white writing area on each end tab for plate identification.
  • Choice of five colours for easy plate identification
  • Applications: Short-term storage, incubation, ELISA
  • Recommended temperature range −40 to +120 °C
End tabs with backing removed adhere to the ends of the plate and are perforated so they can be left in place even if the centre portion of the film is removed. Alternatively, one inscribed colour coded end tab can be removed and pasted in a laboratory notebook while the other remains on the plate.