VWR® Plate Strip Films 2×8, Polyester
Supplier: VWR International
SPS2X8VWR50 SP2X8VWR50
60941-068CS 142.44 CAD
60941-068 60941-066
VWR® Plate Strip Films 2×8, Polyester
Seals Microplate Seals
Polyester 2×8 strip films are identical to polyester films in material and adhesive, but designed for sealing only one or two 8-well rows at a time on either strip-well plates or standard plates whenever rows must be selectively protected or accessed.
  • Seal selectively, one or two rows at a time
  • Can use as secondary seal over previously pierced films
  • Recommended temperature range −40 to +120 °C
Each film L×D: 103,2×19,1 mm, including two removable positioning tabs; long dimension with tabs removed is 82,6 mm.
Packaging: Sterile product is packaged in tamper evident bags of 50/bag.