Omega® Long Graduated Pipette Tips, Cole-Parmer®, Antylia Scientific
Supplier: Antyila Scientific
24501-60 24501-61 24501-65 24501-66 24501-67 24502-43 24502-50 24502-51 24502-52 24502-53 24502-54 24502-81 24502-88 24502-92
76379-946PK 64.53 CAD
76379-946 76379-948 76379-950 76379-952 76379-954 76380-008 76380-010 76380-012 76380-014 76380-016 76380-018 76380-038 76380-040 76380-042
Omega® Long Graduated Pipette Tips, Cole-Parmer®, Antylia Scientific
Pipette Tips
These extended length clear pipette tips crafted of PP feature a special design to prevent DNA denaturalization and adapt with a wide-range of pipettes.
  • Made in a touch-free manufacturing facility with a stringent control process
  • Reach the bottom of vessels without the pipette shaft touching its sides further protecting samples
  • Tip Options: Standard or low retention, sterile, filter and combinations
  • Graduation marks act as visual checks to lessen any risk of error, during sample aspiration
  • Produced from a Swiss-made diamond finish mold
  • Made of FDA approved
More Product Information