VWR® SMS Lab Coats, Made in USA
Supplier: VWR International
76457-004 76457-006 76457-008 76457-010 76457-012 76457-014 76457-016 76457-018 76457-020 76457-022 76457-024 76457-026 76457-028 76457-030 76457-032 76457-034 76457-036 76457-038 76457-040 76457-042 76457-044 76457-046 76457-048 76457-050 76457-052 76457-054 76457-056 76457-058 76457-060 76457-062 76457-064 76457-066 76457-068 76457-070 76457-072 76457-074 76457-076 76457-078 76457-080 76457-082 76457-154 76457-156 76457-158 76457-160 76457-162 76457-164 76457-166 76457-168 76463-370 76463-372 76463-374 76463-376 76470-574 76470-576 76470-578 76470-580 76470-582 76470-584 76470-586 76470-588 76470-590 76470-592 76470-594 76470-596 76470-598 76470-600 76470-602 76470-604 76470-606 76470-608 76470-610 76470-612 76470-614 76470-616 76470-618 76470-620 76470-622 76470-624 76470-626 76470-628
76457-004CS 810.03 CAD
76457-004 76457-006 76457-008 76457-010 76457-012 76457-014 76457-016 76457-018 76457-020 76457-022 76457-024 76457-026 76457-028 76457-030 76457-032 76457-034 76457-036 76457-038 76457-040 76457-042 76457-044 76457-046 76457-048 76457-050 76457-052 76457-054 76457-056 76457-058 76457-060 76457-062 76457-064 76457-066 76457-068 76457-070 76457-072 76457-074 76457-076 76457-078 76457-080 76457-082 76457-154 76457-156 76457-158 76457-160 76457-162 76457-164 76457-166 76457-168 76463-370 76463-372 76463-374 76463-376 76470-574 76470-576 76470-578 76470-580 76470-582 76470-584 76470-586 76470-588 76470-590 76470-592 76470-594 76470-596 76470-598 76470-600 76470-602 76470-604 76470-606 76470-608 76470-610 76470-612 76470-614 76470-616 76470-618 76470-620 76470-622 76470-624 76470-626 76470-628
VWR® SMS Lab Coats, Made in USA
Disposable Garments
The VWR® Made in USA PPE line has been expanded to include SMS lab coats.
  • Multilayer SMS material protection
  • Light fluid and particulate barrier
  • Excellent breathability
  • Serged seams
  • Standard or knit collar options
  • Elastic, open, or knit cuff design
  • White or blue color options
Available in two colors and a variety of styles to meet virtually all disposable lab coat needs. Choose from different cuff options, such as knit, elastic, or no cuff. Additionally, different pocket options are available, including the popular no pocket choice.
Ordering information: Also available in the Made in USA PPE line are masks, coveralls, scrubs, and microporus lab coats.
Packaging: Packed 30 coats per case.