SP Wilmad-LabGlass Precision NMR Suprasil® (Synthetic Quartz) Tubes, SP Industries
Supplier: SP Industries
507-PP-9SUP-5 528-PP-7SUP-5
76459-620PK 942 CAD
76459-620 76459-858
SP Wilmad-LabGlass Precision NMR Suprasil® (Synthetic Quartz) Tubes, SP Industries
Tubes NMR Tubes
Wilmad precision NMR Suprasil® (Synthetic Quartz) tubes are manufactured from high purity synthetic fused silica materials with <0.005 ppm Fe₂O₃ and outstanding optical characteristics in the deep UV to the near IR.
  • Transmission rate from 190 to 2600 nm is well over 95% (10 mm thickness) excluding reflection
  • Possesses a similar thermal expansion rate and tensile strength as natural quartz which is 14 times more robust during the cooling/heating process than Type 1 Class A glass
  • Tight Int.Ø and Ext.Ø tolerance as small as 0.0065 mm accommodates Wilmad
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable