SP Wilmad-LabGlass X Band Capillary Tubes, SP Industries
Supplier: SP Industries
Q-1.5X1.8 Q-.9X1.1 Q-1.0X1.2
76460-074PK 285.4 CAD
76460-074 76460-070 76460-072
SP Wilmad-LabGlass X Band Capillary Tubes, SP Industries
Tubes NMR Tubes
Wilmad's X-band EPR tubes are made from clear fused quartz with the lowest impurities to guarantee success in detecting subtle spectroscopic details.
  • Available with both ends open
  • Ideal for applications with limited sample size
  • Easy to seal with a hot flame or sealant of your choice
Rigorous testing is done on each tube.
Order Now
SPECIFICATIONS
Environmentally Preferable