VWR® A-Series Balances, with NTEP Certificate
Supplier: VWR
VWR®
VWR-4500AC.N VWR-105AC.N VWR-160AC.N VWR-6100AC.N VWR-310AC.N VWR-8100AC.N VWR-220AC.N VWR-205AC.N
76458-458EA 12236.48 CAD
76458-458 76458-468 76458-472 76458-462 76458-470 76458-460 76458-466 76458-464
VWR® A-Series Balances, with NTEP Certificate
Balances and Scales
VWR® A-Series balances incorporate state-of-the-art features for advanced weighing applications and documentation.
  • Internal calibration
  • Multiple weigh modes
  • Voltage: 100 to 240 V
These economical balances offer a variety of standard features, such as internal calibration, GLP procedures, 17 weighing units, adjustable ambient condition settings, 11 application modes, and multilingual operation. The VWR® A-Series balances are the answer for organizations demanding precision and performance at an economical price.
Certifications: NTEP certified.