HM500 Series Half Mask Respirators, Honeywell Safety
Supplier: Honeywell Safety Products
HM501TS HM501TM HM501TL HM502TS HM502TM HM502TL
76452-346CS 156.57 CAD
76452-346 76452-348 76452-350 76452-352 76452-354 76452-356
HM500 Series Half Mask Respirators, Honeywell Safety
Respirators Reusable Respirators Half Face Respirators
Offering breakthrough comfort, and reliable on the job safety, HM500 series half mask respirators are available in threaded/fixed or drop down styles.
  • Soft material construction is comfortable on the skin to promote extended use
  • Lighter than other masks with cartridges in the market
  • Effective seal and good field of vision
  • Easy cleaning and maintenance
  • Low-profile cartridges enable a clear line of sight
  • Drop down option allows for quick work breaks
  • Compatible with all N-Series filters and cartridges
The Honeywell HM500 Series Half Mask is engineered as a lightweight, durable soft half mask with easy operation features. Available in non-drop down (threaded/fixed) and drop down versions, to ensure better comfort and convenience for extended wear. The drop-down option allows workers to remove the masks without taking off hard hats, earmuffs, or eyewear.
Fully compatible with the HM500, Honeywell N-Series filters and cartridges are quick and easy to attach, securely staying in place to provide confidence and safety on the job.