Cryo Shipper Temperature Monitoring Data Logger Kit, ELPRO
Supplier: ELPRO SERVICES, INC.
802351
76457-564KT 1385.03 CAD
76457-564
Cryo Shipper Temperature Monitoring Data Logger Kit, ELPRO
Data Loggers
ELPRO's next generation of INDEPENDENT monitoring solutions for temperature measurement with external sensors. The ideal Cryo shipper data logger kit to simplify the storage and transport of your valuable products. A convenient 'pause alarm' feature allows users to temporarily turn off the alarming e.g. during cryogenic LN2 tank refills, but still maintain traceability. The data logger kit is reconfigurable and reusable for up to three years and are 100% GxP compliant.
  • Digital display with a visual alarm status that is easy to see, with an 'X' or 'check'
  • All settings, including alarm ranges and logging interval, are simple to program and user-configurable via a free configuration tool
  • High/low or multi-alarm levels with user-defined alarm delay
  • Built-in PDF report generation with embedded raw data (no software required)
  • Comes with 2pt NIST calibration certificate (0 °C, −196 °C)
This compliant and economic solution for Cryo Shipper monitoring is ideal for many transport applications. All settings, including alarm ranges and logging interval, are user-configurable via a free configuration tool. Choose from high/low or multi-alarm levels with user-defined alarm delay. Once configured, insert it into any computer USB port to generate a 21 CFR Part 11 Compliant PDF report with no software or download cables required. Additionally, end users have access to on-screen statistics such as min./max./average, current temperature, and alarm status. If necessary, point-by-point data can be accessed via a free downloadable tool that allows additional notation, reporting, and optional export of raw data to Excel.
Certifications: CE marked. The LIBERO CE meets FDA 21 CFR Part 11 and is fully compliant. The LIBERO CE complies with the ISO standard 19005-1 document management for the long term preservation of electronic documents and is 21 CFR Part 11. The CE is delivered with a validation certificate.