Silicon Nitride TEM Window Grids, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
76042-40 76042-41 76042-42 76042-43 76042-44 76042-45 76042-46 76042-47 76042-48 76042-49 76042-50 76042-51 76042-52 76042-53
76439-562PK 1162.26 CAD
76439-562 76439-564 76439-566 76439-568 76439-570 76439-572 76439-574 76439-576 76439-578 76439-580 76439-582 76439-584 76439-586 76439-588
Silicon Nitride TEM Window Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Silicon Nitride TEM Window Grids perform well under harsh lab conditions.
  • Plasma cleanable - can be vigorously plasma cleaned to remove organic contamination
  • Field to field uniformity - less than 0.5 nm variation in film thickness across an entire production log, not just a single window grid
  • Tolerates temperatures above 1000 °C - supports use in environmental TEMs where dynamic processes are observed at high temperatures
  • Withstands harsh conditions - provides an ideal balance of imaging resolution, chemical stability and mechanical strength
  • Incorporates
More Product Information