Vacuum Drying Chambers, VD and VDL Series, Binder
Supplier: Binder
9630-0005 9630-0006 9630-0007 9630-0013 9630-0014 9630-0015 5013-0223 5013-0219 8012-1961 8012-1962 8012-1963 8012-1967 8012-1968 8012-1969
76458-910EA 16382.86 CAD
76458-910 76458-912 76458-914 76458-916 76458-918 76458-920 76458-922 76458-924 76458-938 76458-940 76458-942 76458-944 76458-946 76458-948
Vacuum Drying Chambers, VD and VDL Series, Binder
Ovens Vacuum Ovens
BINDER’s VD series vacuum drying chambers deliver impressive performance through their precise temperature control and gentle drying. The patented expansion rack technology from BINDER also optimizes heat transfer. The shelves can be repositioned for flexible placement and the interior of the chamber is easy to clean.
  • Fast, gentle drying
  • Optimum heat transfer through large thermal conducting plates
  • Safe work thanks to proven safety concept
  • Also option of modular complete system with vacuum pump and pump chamber
  • Controller with digital display of pressure
More Product Information