Avantor® ACE® UltraCore C18, Analytical HPLC/UHPLC Columns
Supplier: Hichrom
CORE-35F-521 CORE-35F-530 CORE-35F-546 CORE-35F-1021 CORE-35F-1030 CORE-35F-1046 CORE-35F-1521 CORE-35F-1530 CORE-35F-1546 CORE-35F-21GD CORE-35F-30GD CORE-35F-46GD
76457-992EA 1216.84 CAD
76457-992 76457-994 76457-996 76457-998 76458-000 76458-002 76458-004 76458-006 76458-008 76458-010 76458-012 76458-014
Avantor® ACE® UltraCore C18, Analytical HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® UltraCore C18 are high performing solid-core (core-shell) particle columns designed for a wide range of LC applications. This densely bonded octadecyl phase can be used to separate acidic, basic and neutral molecules, based on hydrophobic retention mechanisms.
  • High purity, base deactivated silica with excellent reproducibility
  • High efficiency separations comparable with sub-2 µm packings but lower back pressure
  • Ideal for a wide range of analytes differing in hydrophobicity
These stainless steel columns are available in a variety ofMore Product Information