Eisco Porcelain Desiccator Plates
Supplier: Wards
CH0360A CH0360B CH0360C
470328-334EA 6.8 CAD
470328-334 470336-554 470336-556
Eisco Porcelain Desiccator Plates
Desiccators Desiccators, Plates
These desiccator plates can be used in a desiccator to remove moisture from specimen and protect specimen against water vapor in the air.
  • Made from high quality porcelain
  • 15 cm Ø, 3 cm thick
Eisco Labs manufacture thee using state-of-the-art equipment, and follow high standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind.
They are manufactured with only the highest grade, raw materials and according to international quality standards to ensureMore Product Information