Eisco Safety Shield, 3-Panel Polycarbonate
Supplier: Eisco Scientific
CH0659F
470329-600EA 244 CAD
470329-600
Eisco Safety Shield, 3-Panel Polycarbonate
Shields Safety Shields
Polycarbonate safety shield is perfect for counters, desks, reception areas or any other setting where face to face interaction occurs.
  • Made from high quality 4 mm thick polycarbonate
  • Includes 3 adjustable panels for maximum versatility
  • Transparent, provides excellent visual clarity
  • Measures 24" in height
  • Center panel measures 11.8" wide, and panels on either side measure 8.9" wide
  • Can be easily cleaned on a regular basis with common disinfectants
Safety barrier with 3 adjustable panels is made of 4 mm thick high quality, durable polycarbonate. Transparent material allows optimal visual clarity. Barrier is perfect for counters, desks, reception areas or any other setting where face to face interaction occurs.
This is not a sealable barrier, and by no means will protect you from all elements. Functions as a secondary barrier to personal protective equipment in order to provide an extra layer of defense. Does not have anti-fog or anti-microbial properties.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind.
They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.