Eisco Aluminium Dynamometers, Spring Balance
Supplier: Wards
PH0036BAL PH0036CAL PH0036DAL PH0036EAL PH0036FAL PH0036GAL
470328-774EA 18.7 CAD
470328-774 470328-778 470328-782 470328-786 470328-790 470328-794
Eisco Aluminium Dynamometers, Spring Balance
Dynamometers
High resolution, durable aluminium spring balances are a great tool for accurately assessing the weight of an object in both grams and Newton's
  • Premium design with aluminium tube
  • High resolution, easy to read graduations
  • Dual scale measures in grams and Newtons
  • User friendly
  • Useful for a variety of physics and physical science activities
Extremely durable with an aluminium tube, this dynamometer (spring balance) is easy to read, user firendly, and perfect for classroom use.
A dual scale allows measurements to be taken in grams (g) or Newtons (N).