Eisco Economy Dynamometers, Spring Balance
Supplier: EISCO SCIENTIFIC LLC
PH0036B PH0036D PH0036F PH0036G PH0036C PH0036E PH0036A
470328-772EA 12.9 CAD
470328-772 470328-784 470328-776 470328-788 470328-792 470328-770 470328-780
Eisco Economy Dynamometers, Spring Balance
Dynamometers
High resolution, plastic spring balance is a great tool for accurately assessing the weight of an object in both grams and Newton's.
  • Economy grade, plastic tube
  • High resolution, easy to read graduations
  • Dual scale measures in grams and Newtons
  • User friendly
  • Useful for a variety of physics and physical science activities
While economical with a plastic tube this dynamometer (spring balance) is easy to read, user friendly, and perfect for classroom use. A dual scale allows measurements to be taken in grams (g) or Newtons (N).