SensiCare® Polyisoprene Surgical Gloves with Aloe, Medline Industries
Supplier: Medline
MSG1060 MSG1065 MSG1070 MSG1075 MSG1090 MSG1055 MSG1080 MSG1085
54720-111CS 714.37 CAD
54720-111 54720-113 54720-115 54720-117 76332-767 89068-645 89068-647 89068-649
SensiCare® Polyisoprene Surgical Gloves with Aloe, Medline Industries
Gloves
SensiCare® Polyisoprene surgical gloves with aloe contain pure, freeze-dried aloe vera to help soothe and moisturize hands. Healthcare providers must constantly wash their hands and wear gloves as an infection control measure. Repeated glove use and exposure to harsh chemicals can lead to dry, irritated skin. Patented a process to bring the moisturizing properties of aloe vera to surgical gloves.
  • Ideal for general surgery
  • Coated with aloe vera to soothe and moisturize skin
  • Made from unique synthetic polyisoprene, isolex
  • Smooth finish with controlled grip for excellent handling
  • Synthetic polymer coating for effortless damp hand donning
  • ARC (anti roll-down cuff) designed to reduce cuff roll-down common with many gloves
  • Not made with natural rubber latex
Surgeons and staff can feel confident knowing that the surgical gloves produces are 100% inspected for pinholes, tears, and visual defects. 0.65 AQL is 57% more stringent than the FDA requires.
Certifications: Testing meets or exceeds ASTM, EN, and ISO standards.