DuPont™ Tyvek® 1422A, Autoclave Wrappers, Round, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
T2ASHEET06 T2ASHEET10 T2ASHEET14
76451-554CS 4138.53 CAD
76451-554 76451-556 76451-558
DuPont™ Tyvek® 1422A, Autoclave Wrappers, Round, Keystone Cleanroom Products
Covers
Improve the microbial barrier and cleanliness of your sterilization wrap.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Round shaped Tyvek® Wrappers are an ideal way to cover round openings for steam sterilization. Eliminate the wasted time and expense of cutting larger square sheets of sterilization wrap to desired shapes and sizes. Clean cut circles delivered in poly bags ensure cleanliness. Simply secure in place with autoclaveable elastic bands.