DuPont™ Tyvek® Mini-Muff, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
MFT2A03 MFT2A04 MFT2A05 MFT2A05OT MFT2A06BT MFT2A07 MFT2A08 MFT2A09 MFT2A12 MFT2A16 MFT2A18
76450-418CS 5162.56 CAD
76450-418 76450-420 76450-422 76450-424 76450-426 76450-428 76450-430 76450-432 76450-434 76450-436 76450-438
DuPont™ Tyvek® Mini-Muff, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Conveniently packaged-100 per sealed poly bag
Save time and standardize the activities in equipment preparation for sterilization. Tyvek® Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closuresMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable