Mini-Muff Closures, Polypropylene, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
MFNWHD04 MFNWHD05 MFNWHD06 MFNWHD07 MFNWHD08 MFNWHD09 MFNWHD12 MFNWHD16 MFNWHD18
76450-378CS 2346.3 CAD
76450-378 76450-380 76450-382 76450-384 76450-386 76450-388 76450-390 76450-392 76450-394
Mini-Muff Closures, Polypropylene, Keystone Cleanroom Products
Covers
Keep cleaned non-sterile production hoses protected from external sources of contamination.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Tear and puncture resistant
  • Conveniently packaged-100 per sealed poly bag
These polypropylene muffs are a great choice in non-sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Poly bagged in packs of 100 to help ensure cleanliness up until point of use.
Delivery information: Certificates of conformance sent with each shipment.