BHD Muffs with a Skirt Extension, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
MFNWB18SK02 MFNWB18SK02TL
76450-364CS 6517.5 CAD
76450-364 76450-366
BHD Muffs with a Skirt Extension, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - Autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with BioShield paper
  • 2" skirt extension
  • Conveniently packaged-100 per sealed poly bag
Save time and standardize the activities in equipment preparation for sterilization. BHD Muffs with a skirt extension provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilization to reduce time consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings with the addition of a 2" long skirt extension. The skirt extension provides a safe area to remove the muff without indvertently touching the critical surfaces with the fingers.
Delivery information: Certificates of conformance sent with each shipment.