Polypropylene Bowl Covers, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BWLPPW24E BWLPPW26E BWLPPW30E BWLPPW36E BWLPPW42E BWLPPW48E BWLPPW72E
76450-332CS 1381.71 CAD
76450-332 76450-334 76450-336 76450-338 76450-340 76450-342 76450-344
Polypropylene Bowl Covers, Keystone Cleanroom Products
Covers
Keep cleaned non-sterile production equipment surfaces protected from external sources of contamination; or use in process to prevent cross contamination of products.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Tear and puncture resistant
  • Individually poly bagged
These polypropylene bowl covers are a great choice in non-sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover or used 'in-process' to prevent product cross contamination.
Individually poly bagged to ensureMore Product Information