DuPont™ Tyvek® Autoclavable Stopper Bowl Covers, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BWLT2A20 BWLT2A24 BWLT2A26 BWLT2A30 BWLT2A34 BWLT2A36 BWLT2A38 BWLT2A42 BWLT2A46 BWLT2A48 BWLT2A54 BWLT2A56 BWLT2A60 BWLT2A64 BWLT2A72 BWLT2A90 BWLT2A120 BWLT2A144 BWLT2A32
76450-284CS 4953.3 CAD
76450-284 76450-286 76450-288 76450-290 76450-292 76450-294 76450-296 76450-298 76450-300 76450-302 76450-304 76450-306 76450-308 76450-310 76450-312 76450-314 76450-316 76450-318 76468-902
DuPont™ Tyvek® Autoclavable Stopper Bowl Covers, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Individually poly bagged
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Stopper bowl coversMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable