Tyvek® 1073B / PE Steam Sterilization Chevron Peel Pouches, with Steam Indicator, Heat Seal, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BBMGTVKHS0306 BBMGTVKHS0415 BBMGTVKHS0608 BBMGTVKHS0610 BBMGTVKHS0612 BBMGTVKHS0810 BBMGTVKHS0815 BBMGTVKHS1020 BBMGTVKHS1030 BBMGTVKHS1212 BBMGTVKHS1217 BBMGTVKHS1218 BBMGTVKHS1224 BBMGTVKHS1617 BBMGTVKHS1822 BBTMGTVK1840 BBMGTVKHS2418 BBMGTVKHS2424 BBMGTVKHS2430 BBTMGTVK2442 BBTMGTVK2440 BBTMGTVK2448 BBTMGTVK12-18 BBTMGTVK12-30 BBMGTVKHS1624 BBMGTVKHS1626 BBMGTVKHS1630 BBMGTVKHS1632 BBMGTVKHS1635 BBMGTVKHS1816 BBMGTVKHS2426 BBMGTVKHS2435 BBMGTVKHS0632 BBMGTVKHS0635 BBMGTVKHS0710 BBMGTVKHS0713 BBMGTVKHS0807 BBMGTVKHS0814 BBMGTVKHS0818 BBMGTVKHS0834 BBMGTVKHS1011 BBMGTVKHS1012 BBMGTVKHS0206 BBMGTVKHS0208 BBMGTVKHS02095 BBMGTVKHS0308 BBMGTVKHS03508 BBMGTVKHS0422 BBMGTVKHS0510 BBMGTVKHS0620 BBMGTVKHS0625 BBMGTVKHS0628 BBMGTVKHS1015 BBMGTVKHS1214 BBMGTVKHS1220 BBMGTVKHS1420 BBMGTVKHS1424 BBMGTVKHS1516 BBMGTVKHS1520 BBMGTVKHS1616 BBMGTVKHS1618 BBMGTVKHS1620
76450-826CS 1482.44 CAD
76450-826 76450-828 76450-830 76450-832 76450-834 76450-836 76450-838 76450-840 76450-842 76450-844 76450-846 76450-848 76450-850 76450-852 76450-854 76450-856 76450-858 76450-860 76450-862 76450-864 76450-866 76450-868 76637-342 76637-344 77335-344 77335-346 77335-348 77335-350 77335-352 77335-354 77335-356 77335-358 77335-304 77335-306 77335-308 77335-310 77335-312 77335-314 77335-316 77335-318 77335-320 77335-322 77335-284 77335-286 77335-288 77335-290 77335-292 77335-294 77335-296 77335-298 77335-300 77335-302 77335-324 77335-326 77335-328 77335-330 77335-332 77335-334 77335-336 77335-338 77335-340 77335-342
Tyvek® 1073B / PE Steam Sterilization Chevron Peel Pouches, with Steam Indicator, Heat Seal, Keystone Cleanroom Products
Bags Sterilization Pouches
Improve the performance, microbial barrier and cleanliness of your autoclave peel pouches.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Peels open cleanly
  • Chevron peel
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Heat seals closed
  • Integrated steam sterilization indicator
A significant upgrade over paper-based steam sterilization pouches. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinely do.
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
Delivery information: Certificates of Conformance sent with each shipment.