Tyvek® 1073B Autoclave Bags with Steam Indicator, Heat Seal, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
COVMGTVKUB1X14 COVMGTVKUB3X6 COVMGTVKUB3X36 COVMGTVKUB5X25 COVMGTVKUB6X8 COVMGTVKUB6X16 COVMGTVKUB6X22 COVMGTVKUB6X34 COVMGTVKUB8X9 COVMGTVKUB8X12 COVMGTVKUB8X14 COVMGTVKUB8X30 COVMGTVK10X12 COVMGTVK10X20 COVMGTVK10X25 COVMGTVK12X12 COVMGTVK12X16 COVMGTVK12X18 COVMGTVK12X24 COVMGTVK12X32 COVMGTVK12X34 COVMGTVK14X22 COVMGTVK14X28 COVMGTVK14X42 COVMGTVK15X18 COVMGTVK16X24 COVMGTVK16X32 COVMGTVK16X36 COVMGTVK16X44 COVMGTVK18X22 COVMGTVK18X24 COVMGTVK18X26 COVMGTVK18X30 COVMGTVK18X40 COVMGTVK18X68 COVMGTVK20X20 COVMGTVK20X32 COVMGTVK24X24 COVMGTVK24X44 COVMGTVK25X37 COVMGTVK26X28 COVMGTVK26X30 COVMGTVK30X30 COVMGTVK30X22 COVMGTVK30X32 COVMGTVK32X34 COVMGTVK34X34 COVMGTVK34X40 COVMGTVK36X36 COVMGTVK36X42 COVMGTVK36X48 COVMGTVK36X60 COVMGTVK38X40 COVMGTVK38X48 COVMGTVK40X48 COVMGTVK42X46 COVMGTVK48X28 COVMGTVK48X48 COVMGTVK48X64
76450-514CS 667.42 CAD
76450-514 76450-516 76450-518 76450-520 76450-522 76450-524 76450-526 76450-528 76450-530 76450-532 76450-534 76450-536 76450-538 76450-540 76450-542 76450-544 76450-546 76450-548 76450-550 76450-552 76450-554 76450-556 76450-558 76450-560 76450-562 76450-564 76450-566 76450-568 76450-570 76450-572 76450-574 76450-576 76450-578 76450-580 76450-582 76450-584 76450-586 76450-588 76450-590 76450-592 76450-594 76450-596 76450-598 76450-600 76450-602 76450-604 76450-606 76450-608 76450-610 76450-612 76450-614 76450-616 76450-618 76450-620 76450-622 76450-624 76450-626 76450-628 76450-630
Tyvek® 1073B Autoclave Bags with Steam Indicator, Heat Seal, Keystone Cleanroom Products
Bags Sterilization Pouches
1073B Tyvek® are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps as well as heavy equipment parts with sharp edges.
  • Manufactured and packaged in a controlled cleanroom environment
  • Steam sterilization indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic and lint-free
  • 100% Medical Grade 1073B Tyvek® permits fast steam penetration
  • Excellent impact resistance with exceptional microbial barrier
  • Consistently strong factory seals
100% Medical Grade 1073B Tyvek® Autoclave Bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps.
With consistently strong factory seals, these are a good choice for heavy, or pharmaceutical change parts with sharp corners.
All Tyvek construction allows for a greater number of bags per sterilization load; increasing efficiencies and cost reduction.
Delivery information: Certificates of Conformance sent with each shipment.