Tyvek® 1073B PE Steam Sterilization Tubing, Continuous Form, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BBTMGTVK04 BBTMGTVK06 BBTMGTVK09 BBTMGTVK12 BBTMGTVK14 BBTMGTVK18 BBTMGTVK24
76451-000CS 1598.2 CAD
76451-000 76451-002 76451-004 76451-006 76451-008 76451-010 76451-012
Tyvek® 1073B PE Steam Sterilization Tubing, Continuous Form, Keystone Cleanroom Products
Bags Sterilization Pouches
Improve the prep time, performance, microbial barrier and cleanliness of your autoclave peel pouches.
  • Manufactured and packaged in a controlled cleanroom environment
  • Sizing flexibility and hydrophobic
  • Plastic core (no corrugated material)
  • Unroll and cut to desired length, heat seal both ends
  • Autoclavable at 121 to 125 °C
  • Peels open cleanly, lint-free
  • Tear and puncture resistant
A significant upgrade over paper based steam sterilization tubing . These Tyvek® peel pouches will not rip or puncture like paper based autoclave pouches routinely do. TheMore Product Information