Tyvek® 1422A Autoclave Bags with Steam Indicator, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BGT2A0.7505 BGT2A0.7508 BGT2A0105 BGT2A0106 BGT2A0108 BGT2A0205 BGT2A0306 BGT2A0404 BGT2A0406 BGT2A0409 BGT2A0416 BGT2A0430 BGT2A0610 BGT2A0612 BGT2A0638 BGT2A0820 BGT2A09512 BGT2A1015 BGT2A1030 BGT2A1062 BGT2A1212 BGT2A1216 BGT2A1218 BGT2A1232 BGT2A1234 BGT2A1240 BGT2A1246 BGT2A1248 BGT2A1416 BGT2A1418 BGT2A1424 BGT2A1543 BGT2A1604 BGT2A1612 BGT2A1624 BGT2A1638 BGT2A1824 BGT2A1830 BGT2A1836 BGT2A2020 BGT2A2024 BGT2A2030 BGT2A2040 BGT2A2224 BGT2A2424 BGT2A2436 BGT2A2442 BGT2A2448 BGT2A2462 BGT2A2628 BGT2A2844 BGT2A3016 BGT2A3020 BGT2A3036 BGT2A3040 BGT2A3060 BGT2A3412 BGT2A3636 BGT2A3642 BGT2A3648 BGT2A3660 BGT2A4242 BGT2A4416 BGT2A4848 BGT2A4864 BGT2A5054 BGT2A2472 BGT2A2818 BGT2A2834 BGT2A2838 BGT2A2460 BGT2A2840 BGT2A2864 BGT2A3030 BGT2A3246 BGT2A3434 BGT2A4040 BGT2A4836 BGT2A4860 BGT2A4872 BGT2A5050 BGT2A0124 BGT2A0203 BGT2A0208 BGT2A0236 BGT2A0250 BGT2A0309 BGT2A0310 BGT2A0412 BGT2A0448 BGT2A0516 BGT2A0522 BGT2A0526 BGT2A0608 BGT2A0662 BGT2A0710 BGT2A0808 BGT2A0809 BGT2A0812 BGT2A0816 BGT2A0822 BGT2A0824 BGT2A0856 BGT2A0872 BGT2A1010 BGT2A1020 BGT2A1024 BGT2A1222 BGT2A1224 BGT2A1236 BGT2A1252 BGT2A1420 BGT2A1422 BGT2A1426 BGT2A1526 BGT2A1530 BGT2A1534 BGT2A1620 BGT2A1632 BGT2A1818 BGT2A1848 BGT2A1855 BGT2A2026 BGT2A2028 BGT2A2044 BGT2A2222 BGT2A2225 BGT2A2226 BGT2A2325 BGT2A2430 BGT2A2440 BGT2A506 BGT2A606
76451-344CS 4034.77 CAD
76451-344 76451-346 76451-348 76451-350 76451-352 76451-354 76451-356 76451-358 76451-360 76451-362 76451-364 76451-366 76451-368 76451-370 76451-372 76451-374 76451-376 76451-378 76451-380 76451-382 76451-384 76451-386 76451-388 76451-390 76451-392 76451-394 76451-396 76451-398 76451-400 76451-402 76451-404 76451-406 76451-408 76451-410 76451-412 76451-414 76451-416 76451-418 76451-420 76451-422 76451-424 76451-426 76451-428 76451-430 76451-432 76451-434 76451-436 76451-438 76451-440 76451-442 76451-444 76451-446 76451-448 76451-450 76451-452 76451-454 76451-456 76451-458 76451-460 76451-462 76451-464 76451-466 76451-468 76451-470 76451-472 76451-474 76468-870 76468-872 76468-874 76468-876 76468-878 76468-880 76468-882 76468-884 76468-886 76468-888 76468-890 76468-892 76468-894 76468-896 76468-898 76469-820 76469-822 76469-824 76469-826 76469-828 76469-830 76469-832 76469-834 76469-836 76469-838 76469-840 76469-842 76469-844 76469-846 76469-848 76469-850 76469-852 76469-854 76469-856 76469-858 76469-860 76469-862 76469-864 76469-866 76469-868 76469-870 76469-872 76469-874 76469-876 76469-878 76469-880 76469-882 76469-884 76469-886 76469-888 76469-890 76469-892 76469-894 76469-896 76469-898 76469-900 76469-902 76469-904 76469-906 76469-908 76469-910 76469-912 76469-914 76469-916 76469-918 76469-920 76469-922
Tyvek® 1422A Autoclave Bags with Steam Indicator, Keystone Cleanroom Products
Bags Sterilization Pouches
Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
  • Tear and puncture resistant
  • Designed to be heat sealed closed
  • Superior to paper based autoclave pouches and wraps
  • Fast and consistent for equipment preparation area
  • Steam indicator and hydrophobic
  • Lint-free; strong factory seals
  • Exceptional microbial barrier
Standardize and simplify the way equipment is wrapped for steam sterilization. The use of these bags will also save valuable time in the equipment prep area.
The drapeable nature of the 1422A Tyvek® material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficieny and throughput.
Manufactured and packaged in a controlled cleanroom environment.
Delivery information: Supplied with Certificate of Conformance.