Tyvek® Stopper Bag Tubings, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
SBTMGTVK02 SBTMGTVK03 SBTMGTVK04 SBTMGTVK06 SBTMGTVK09 SBTMGTVK10 SBTMGTVK12 SBTMGTVK18 SBTMGTVK28
76450-496CS 716.93 CAD
76450-496 76450-498 76450-500 76450-502 76450-504 76450-506 76450-508 76450-510 76450-512
Tyvek® Stopper Bag Tubings, Keystone Cleanroom Products
Bags Sterilization Pouches
1073B Tyvek® / HDPE autoclave continuous form tubing is ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps. Also a good choice for heavy equipment parts where visibility into the bag is required. Tubing form allows for excellent sizing flexibility.
  • Manufactured and packaged in a controlled cleanroom environment
  • Steam sterilization indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic and lint-free
  • Semi-transparent HDPE film side
  • Excellent impact resistance with exceptional microbial barrier
  • Consistently strong, double welded factory seals
1073B Tyvek® side permits steam penetration during sterilization. These are a good choice for heavy, or pharmaceutical change parts with sharp corners or where visibility into the bag is required after sterilization.
Simply unroll the tube to the desired length, heat seal one end to create the bag form, add components to the bag then heat seal the open end.
Certifications: Supplied with Certificates of conformance.