GMP Covers, Grey Tyvek® Cylidrical Shaped with Adjustable Tyvek Drawstring Closure, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
COVGTVK6012D
76451-774CS 6860.42 CAD
76451-774
GMP Covers, Grey Tyvek® Cylidrical Shaped with Adjustable Tyvek Drawstring Closure, Keystone Cleanroom Products
Covers
These Grey Tyvek covers block UV light, which can damage light sensitive biotech products. Use in any pharmaceutical manufacturing setting to keep cleaned equipment protected from contamination.
  • Manufactured and packaged in a controlled cleanroom environment
  • Blocks UV Light
  • Reduces labor costs
  • Simplifies the conformance to SOP's
  • Individually poly bagged and labeled
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Grey UV Light blocking Tyvek Covers can be used in all pharmaceutical manufacturing environments to helps minimize microbial and particulate contamination risks.
Certifications: Certificates of Conformance sent with each shipment.