Tyvek® Stopper Bags, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
SBMGTVKHS0304 SBMGTVKHS0306 SBMGTVKHS0409 SBMGTVKHS0422 SBMGTVKHS0508 SBMGTVKHS0610 SBMGTVKHS0630 SBMGTVKHS0812 SBMGTVKHS1035 SBMGTVKHS1212 SBMGTVKHS1215 SBMGTVKHS1219 SBMGTVKHS1238 SBMGTVKHS1318 SBMGTVKHS1414 SBMGTVKHS1518 SBMGTVKHS1616 SBMGTVKHS1632 SBMGTVKHS1824 SBMGTVKHS1920 SBMGTVKHS2022 SBMGTVKHS2026 SBMGTVKHS2216 SBMGTVKHS2418 SBMGTVKHS2436 SBMGTVKHS2440 SBMGTVKHS2848
76450-440CS 1633.96 CAD
76450-440 76450-442 76450-444 76450-446 76450-448 76450-450 76450-452 76450-454 76450-456 76450-458 76450-460 76450-462 76450-464 76450-466 76450-468 76450-470 76450-472 76450-474 76450-476 76450-478 76450-480 76450-482 76450-484 76450-486 76450-488 76450-490 76450-492
Tyvek® Stopper Bags, Keystone Cleanroom Products
Bags Sterilization Pouches
1073B Tyvek® / HDPE autoclave bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps. Also a good choice for heavy equipment parts where visibility into the bag is required.
  • Manufactured and packaged in a controlled cleanroom environment
  • Steam sterilization indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic and lint-free
  • Semi-transparent HDPE film side
  • Excellent impact resistance with exceptional microbial barrier
  • Consistently strong, double welded factory seals
Order Now
SPECIFICATIONS
Environmentally Preferable