Cleanroom Facemasks, Gamma Irradiated, Green, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
S1461XLDEL2AD S1461DEL2AD 1461ELAD20PB S1461AD S1461XLGAD S1461ELAD 1461AD20PB
76451-890CS 3613.86 CAD
76451-890 76451-888 76451-874 76451-884 76451-882 76451-870 76451-880
Cleanroom Facemasks, Gamma Irradiated, Green, Keystone Cleanroom Products
Masks Cleanroom Masks
These masks are with no-fog adhesive strip.
Certifications: Certificates of irradiation and conformance with each shipment.