BHD Autoclave Bags with Self-Seal Closure, Blue, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BNW0638SS2P BNW0840SS2P BNW0844SS2P BNW1240SS2P BNW1622SS2P BNW1648SS2P BNW2226SS2P BNW2626SS2P BNW2646SS2P BNW2832SS2P BNW4040SS2P BNW4446SS2P BNW4448SS2P BNW5858SS2P
76451-526CS 3163.05 CAD
76451-526 76451-528 76451-530 76451-532 76451-534 76451-536 76451-538 76451-540 76451-542 76451-544 76451-546 76451-548 76451-550 76451-552
BHD Autoclave Bags with Self-Seal Closure, Blue, Keystone Cleanroom Products
Bags Sterilization Pouches
Self-sealing BHD Autoclave bags can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with BioShield paper
  • Eliminates need to cut material and the use of autoclave tape
Standardize and simplify the way equipment is wrapped for steam sterilization.
The use of these 2More Product Information