BHD Autoclave Bags with Tyvek® Drawstring Closure, Blue, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BNWBG0606D2P BNWBG0642D2P BNWBG09.512D2P BNWBG1212D2P BNWBG1218D2P BNWBG1624D2P BNWBG01644D2P BNWBG1830D2P BNWBG1854D2P BNWBG2024D2P BNWBG2032D2P BNWBG2424D2P BNWBG2435D2P BNWBG2436D2P BNWBG3648D2P BNWT3654D22P BNWBG4040D2P
76451-492CS 8558.32 CAD
76451-492 76451-494 76451-496 76451-498 76451-500 76451-502 76451-504 76451-506 76451-508 76451-510 76451-512 76451-514 76451-516 76451-518 76451-520 76451-522 76451-524
BHD Autoclave Bags with Tyvek® Drawstring Closure, Blue, Keystone Cleanroom Products
Bags Sterilization Pouches
BHD Autoclave bags with Tyvek® Drawstrings can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with BioShield paper
  • Eliminates need to cut material and the use of autoclave tape
Standardize and simplify the way equipment is wrapped forMore Product Information