Tyvek® 1422A Filling Needle Autoclave Bags with Drawstring, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BGT2A0101D BGT2A0102D BGT2A0103D BGT2A0103D10 BGT2A0104D BGT2A0105D BGT2A0106D BGT2A0107D BGT2A0108D BGT2A0111D BGT2A0202D BGT2A0202D16 BGT2A0203D BGT2A0204D BGT2A0205D BGT2A0206D BGT2A0207D BGT2A0208D BGT2A0209D BGT2A0210D BGT2A0211D BGT2A0212D BGT2A0213D BGT2A02503D BGT2A0303D18 BGT2A0304D BGT2A0305D BGT2A0306D BGT2A0308D BGT2A0309D BGT2A0310D BGT2A0312D BGT2A0314D BGT2A0403D BGT2A0404D BGT2A0410D
76451-042CS 3515.53 CAD
76451-042 76451-044 76451-046 76451-048 76451-050 76451-052 76451-054 76451-056 76451-058 76451-060 76451-062 76451-064 76451-066 76451-068 76451-070 76451-072 76451-074 76451-076 76451-078 76451-080 76451-082 76451-084 76451-086 76451-088 76451-090 76451-092 76451-094 76451-096 76451-098 76451-100 76451-102 76451-104 76451-106 76451-108 76451-110 76451-112
Tyvek® 1422A Filling Needle Autoclave Bags with Drawstring, Keystone Cleanroom Products
Bags Sterilization Pouches
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. Also creates secondary level of protection during aseptic presentation.
  • Tear and puncture resistant
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Steam indicator
  • Hydrophobic and lint-free
  • Reliable factory seals
  • Exceptional microbial barrier
These Tyvek® filling needle bags stay in place via the adjustable drawstring closure system. The bags can be aseptically removed with forceps after the filling needles are set in place, without having to cut, or remove particulate generating autoclave tape. These are manufactured and packaged in a controlled cleanroom environment.
Delivery information: Certificates of Conformance sent with each shipment.