BHD Filling Needle Autoclave Bags with Tyvek® Drawstring Closure, Blue, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BNWBG0104D2P BNWBG0106D2P BNWBG0108D2P BNWBG0204D2P BNWBG0206D2P BNWBG0210D2P BNWBG0308D2P BNWBG0310D2P
76451-476CS 7541.2 CAD
76451-476 76451-478 76451-480 76451-482 76451-484 76451-486 76451-488 76451-490
BHD Filling Needle Autoclave Bags with Tyvek® Drawstring Closure, Blue, Keystone Cleanroom Products
Bags Sterilization Pouches
These bags protects needles from accidental touching after they are removed from the primary sterilization packaging.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • 2-ply and steam sterilization indicator - autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with BioShield paper
  • Eliminates need to cut material and the use of autoclave tape
These BHD filling needle bags stay in place via the adjustable Tyvek® drawstring closure system. The bags can be aseptically removed with forceps after the filling needles are set in place, without having to cut, or remove particulate generating autoclave tape.
Certifications: Certificates of Conformance sent with each shipment.