Tyvek® 1422A Steam Sterilization Tubing, Continuous Form, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
TUBT2A02 TUBT2A03 TUBT2A04 TUBT2A05 TUBT2A06 TUBT2A08 TUBT2A10 TUBT2A12 TUBT2A15 TUBT2A16 TUBT2A24 TUBT2A28 TUBT2A30 TUBT2A40
76451-014CS 1629.38 CAD
76451-014 76451-016 76451-018 76451-020 76451-022 76451-024 76451-026 76451-028 76451-030 76451-032 76451-034 76451-036 76451-038 76451-040
Tyvek® 1422A Steam Sterilization Tubing, Continuous Form, Keystone Cleanroom Products
Bags Sterilization Pouches
Continuous Form tubing made from 1422A Tyvek® provides the user with tremendous sizing flexibility when wrapping pharmaceutical equipment for steam sterilization.
  • Manufactured and packaged in a controlled cleanroom environment
  • On a plastic core (no corrugated material)
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Consistently strong factory seals
  • Exceptional microbial barrier
This steam sterilization tubing is a great choice when there is a wide range of equipment parts that need to be wrapped for sterilization.
Simply unroll the tubing to theMore Product Information