DuPont™ Tyvek® 1422A Autoclave Wrappers, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
T2ASHEET0303 T2ASHEET0404 T2ASHEET0606 T2ASHEET0612 T2ASHEET075075 T2ASHEET0808 T2ASHEET0909 T2ASHEET0911 T2ASHEET1212 T2ASHEET1515 T2ASHEET1818 T2ASHEET2020 T2ASHEET2424 T2ASHEET3030 T2ASHEET3636 T2ASHEET4040 T2ASHEET4080 T2ASHEET5454 T2ASHEET60140 T2ASHEET6060 T2ASHEET7272 T2ASHEET8494
76451-560CS 592.53 CAD
76451-560 76451-562 76451-564 76451-566 76451-568 76451-570 76451-572 76451-574 76451-576 76451-578 76451-580 76451-582 76451-584 76451-586 76451-588 76451-590 76451-592 76451-594 76451-596 76451-598 76451-600 76451-602
DuPont™ Tyvek® 1422A Autoclave Wrappers, Keystone Cleanroom Products
Sterilization Wraps
Improve the microbial barrier and cleanliness of your sterilization wrap.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Drapeable
  • Hydrophobic
  • Lint-free
Drapeable 1422A Tyvek® Wrappers are an improvement over cellulose or SMS based wrapping materials for steam sterilization. Eliminate the wasted time and expense of cutting larger square sheets of sterilization wrap to your desired sizes. Clean cut squares and rectangles are delivered in labeled, sealed poly bags to ensure cleanliness.
Certifications: Certificates of conformance sent with each shipment.