DuPont™ Tyvek® 1422A Elasticized Covers for Steam Sterilization, Rectangular Shaped, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
T2ASHEET0324E T2ASHEET1818E T2ASHEET2424E T2ASHEET2440E T2ASHEET2626E T2ASHEET2816E T2SHET2838E2D2 T2ASHEET3014E T2ASHEET3024E T2ASHEET3434E T2ASHEET4080E T2ASHEET4233E T2ASHEET4320E
76451-604CS 3584.63 CAD
76451-604 76451-606 76451-608 76451-610 76451-612 76451-614 76451-616 76451-618 76451-620 76451-622 76451-624 76451-626 76451-628
DuPont™ Tyvek® 1422A Elasticized Covers for Steam Sterilization, Rectangular Shaped, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave the covers in place during installation to prevent microbial and particulate contamination. Leave in place while the remaining fill line set-up activities are performed.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam Sterilization Indicator - Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Elasticized Tyvek® Covers provide a convenient, time-More Product Information