Avantor® ACE® UltraCore BIO Phenyl2, Analytical HPLC/UHPLC Columns
Supplier: Hichrom
BIO-252-521G BIO-252-0521 BIO-252-1021 BIO-252-1521 BIO-252-530G BIO-252-0530 BIO-252-1030 BIO-252-1530 BIO-252-546G BIO-252-0546 BIO-252-1046 BIO-252-1546 BIO-352-521G BIO-352-0521 BIO-352-1021 BIO-352-1521 BIO-352-530G BIO-352-0530 BIO-352-1030 BIO-352-1530 BIO-352-546G BIO-352-0546 BIO-352-1046 BIO-352-1546
76457-850EA 1367.38 CAD
76457-850 76457-856 76457-862 76457-868 76457-874 76457-880 76457-886 76457-892 76457-898 76457-904 76457-910 76457-916 76457-922 76457-928 76457-934 76457-940 76457-946 76457-952 76457-958 76457-964 76457-970 76457-976 76457-982 76457-988
Avantor® ACE® UltraCore BIO Phenyl2, Analytical HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® UltraCore BIO Phenyl2 are high performing solid-core (core-shell) particle columns, designed for a range of biomolecule applications. With both 300 and 500 Å pore sizes available, optimum pore size can be chosen. This phase is well suited to the analysis of hydrophobic proteins over 5kDa molecular weight with alternative selectivity to C4 bonded phases.
  • Wide pore, solid-core combination for excellent peak shape and resolution of large molecules
  • High purity, base deactivated silica with excellent reproducibility
  • Alternative selectivity to
More Product Information