Disodium hydrogen phosphate heptahydrate USP, FCC, Ph. Eur.

Supplier: Macco Organiques S.R.O

51115070120 51115070500
89523-444EA 4066.68 CAD
89523-444 10026-130
Disodium hydrogen phosphate heptahydrate USP, FCC, Ph. Eur.
di-Sodium hydrogen phosphate heptahydrate

For excipient use only.


Certifications: Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.


Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.

Formula: HNa₂O₄P·7H₂O
MDL Number: MFCD00149180
CAS Number: 7782-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR