Ultrathin Carbon Films, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
HC200-AU-CC HC200-AU-FCF HC200-AU-FF HC200-CU-CC HC200-CU-FCF HC200-CU-FF HC200-NI-CC HC200-NI-FCF HC200-NI-FF HC300-AU-CC HC300-AU-FCF HC300-AU-FF HC300-CU-CC HC300-CU-FCF HC300-CU-FF HC300-NI-CC HC300-NI-FCF HC300-NI-FF HC400-AU-CC HC400-AU-FCF HC400-AU-FF HC400-CU-CC HC400-CU-FCF HC400-CU-FF HC400-NI-CC HC400-NI-FCF HC400-NI-FF
76439-746BX 509.58 CAD
76439-746 76439-748 76439-750 76439-754 76439-756 76439-758 76439-762 76439-764 76439-766 76439-772 76439-774 76439-776 76439-780 76439-782 76439-784 76439-788 76439-790 76439-792 76439-796 76439-798 76439-800 76439-804 76439-806 76439-808 76439-812 76439-814 76439-816
Ultrathin Carbon Films, Electron Microscopy Sciences
Graticules, Reticles and Grids
The continuous ultrathin film on holey carbon film allows for the thinnest support film that still has adequate strength to withstand specimen preparation.
  • Ideal for imaging nanotubes, virus particles and other small particulate material
  • Size of the holes ranges from 0.25 to 5 µm
  • TEM imaging with practically no interference from the carbon film supporting it
The film (less than 3 nm thick) lies across a holey carbon film supported by a 200, 300, or 400 mesh grid. The size of the holes differ widely from batch to batch but are generally in the range of ¹/₄ to 5 µm, which gives the equivalent support of at least 6000 mesh grid. Specimen material lying over the covered holes can be imaged in the TEM with practically no interference from the carbon film.