Ultrathin Carbon Films, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
LC200-AU-CC LC200-AU-CC-25 LC200-AU-FCF LC200-AU-FCF25 LC200-AU-FF LC200-AU-FF-25 LC200-CU-CC LC200-CU-CC-25 LC200-CU-FCF LC200-CU-FCF25 LC200-CU-FF LC200-CU-FF-25 LC200-NI-CC LC200-NI-CC-25 LC200-NI-FCF LC200-NI-FCF25 LC200-NI-FF LC200-NI-FF-25 LC300-AU-CC LC300-AU-FCF LC300-AU-FF LC300-CU-CC LC300-CU-FCF LC300-CU-FF LC305-NI-CC LC305-NI-FCF LC305-NI-FF LC325-AU-CC LC325-AU-FCF LC325-AU-FF LC325-CU-CC LC325-CU-FCF LC325-CU-FF LC325-NI-CC LC325-NI-FCF LC325-NI-FF LC400-AU-CC LC400-AU-CC-25 LC400-AU-FCF LC400-AU-FCF25 LC400-AU-FF LC400-AU-FF-25 LC400-CU-CC LC400-CU-CC-25 LC400-CU-FCF LC400-CU-FCF25 LC400-CU-FF LC400-CU-FF-25 LC400-NI-CC LC400-NI-CC-25 LC400-NI-FCF LC400-NI-FCF25 LC400-NI-FF LC400-NI-FF-25
76439-822BX 82.84 CAD
76439-822 76439-824 76439-826 76439-828 76439-830 76439-832 76439-836 76439-838 76439-840 76439-842 76439-844 76439-846 76439-850 76439-852 76439-854 76439-856 76439-858 76439-860 76439-864 76439-866 76439-868 76439-872 76439-874 76439-876 76439-882 76439-884 76439-886 76439-888 76439-890 76439-892 76439-894 76439-896 76439-898 76439-900 76439-902 76439-904 76439-906 76439-908 76439-910 76439-912 76439-914 76439-916 76439-920 76439-922 76439-924 76439-926 76439-928 76439-930 76439-934 76439-936 76439-938 76439-940 76439-942 76439-944
Ultrathin Carbon Films, Electron Microscopy Sciences
Graticules, Reticles and Grids
The continuous ultrathin film on lacey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. The film (less than 3 nm thick) lies across a carbon lacey film supported by a 200, 300, or 400 mesh grid. This product is ideal for looking at nanotubes, virus particles and other small particulate material.
  • Lacey carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes
The size of the holes in the lacey film differ widely from batch to batch but are generally in the range of ¹/₄ to 5 µm, which gives the equivalent support of at least 6000 mesh grid. Specimen material lying over the covered holes can be imaged in the TEM with practically no interference from the carbon film.