Tomography Grids, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
74357 74357-01 74357-02 74357-03 74357-04
76439-534PK 362.26 CAD
76439-534 76439-536 76439-538 76439-540 76439-542
Tomography Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
This 1.5 mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90°.