Picking Trays, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
71171-01 71171-02 71171-03
76439-514EA 54.84 CAD
76439-514 76439-516 76439-518
Picking Trays, Electron Microscopy Sciences
Grid Holders
Trays perfect for placement of grids and storage of small components. The trays feature 42 cells with a lip differentiating each cell. The cells measure 11×11 mm (⁷/₁₆×⁷/₁₆").
These unique trays are available in either black rigid ABS plastic or aluminum in black or tan.