mPrep™ Pipettor Kits, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
85010-07 85010-08
76439-626EA 1478.25 CAD
76439-626 76439-628 76439-630 76439-632
mPrep™ Pipettor Kits, Electron Microscopy Sciences
Pipettes Micropipettes
Choose from either single- or multi-channel pipettor kits built around mPrep/g™ capsules.
Single-channel kit includes single channel 200 µl pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips, size 10 µl.
Eight-channel kit includes eight-channel 200 µl pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips, size 10 µl.