EMS Grid Prep Holders, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
71175-00 71175-01 71175-02
76439-520EA 445.42 CAD
76439-520 76439-522 76439-524
EMS Grid Prep Holders, Electron Microscopy Sciences
Grid Holders
EMS preparation tray may be use for staining and/or placing an evaporator for the coating of grids.
  • Medical grade aluminum
  • Includes a plastic storage box
These unique trays have a M4 tapped hole in the center which allows for insertion of pin mount. They also have a screw pin in the middle that may be used as a handle for manipulating the trays.